Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAATATTAAATTGTTGCACCTTAC[A/G]AACAGTGGAAAAGAAAAAAAAAGTA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 600922 | |||||||||||||||||||||||
Literature Links: |
MYLK PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
MYLK - myosin light chain kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321309.1 | 7329 | UTR 3 | NP_001308238.1 | |||
NM_053025.3 | 7329 | UTR 3 | NP_444253.3 | |||
NM_053026.3 | 7329 | UTR 3 | NP_444254.3 | |||
NM_053027.3 | 7329 | UTR 3 | NP_444255.3 | |||
NM_053028.3 | 7329 | UTR 3 | NP_444256.3 | |||
XM_011512860.2 | 7329 | Intron | XP_011511162.1 | |||
XM_011512861.2 | 7329 | Intron | XP_011511163.1 | |||
XM_017006469.1 | 7329 | Intron | XP_016861958.1 | |||
XM_017006470.1 | 7329 | Intron | XP_016861959.1 | |||
XM_017006471.1 | 7329 | Intron | XP_016861960.1 | |||
XM_017006472.1 | 7329 | UTR 3 | XP_016861961.1 | |||
XM_017006473.1 | 7329 | UTR 3 | XP_016861962.1 |
MYLK-AS1 - MYLK antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |