Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTTGAAGGCCAAGGATCACAGAA[A/G]CATTTATCCTTCTTTTCTCACATGT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 610124 | |||||||||||||||||||||||
Literature Links: |
C3orf22 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
C3orf22 - chromosome 3 open reading frame 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152533.2 | Intron | NP_689746.1 | ||||
XM_011512455.2 | Intron | XP_011510757.1 | ||||
XM_011512457.2 | Intron | XP_011510759.1 | ||||
XM_017005751.1 | Intron | XP_016861240.1 | ||||
XM_017005752.1 | Intron | XP_016861241.1 | ||||
XM_017005753.1 | Intron | XP_016861242.1 |
CHST13 - carbohydrate sulfotransferase 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152889.2 | Intron | NP_690849.1 |