Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGACTCTTGTAGCTCATTAACTA[A/G]TTTTCAACTGCTTTGCTCCTCTCCC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 614658 | |||||||||||||||||||||||
Literature Links: |
AMOTL2 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
AMOTL2 - angiomotin like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278683.1 | 3623 | UTR 3 | NP_001265612.1 | |||
NM_001278685.1 | 3623 | UTR 3 | NP_001265614.1 | |||
NM_016201.3 | 3623 | UTR 3 | NP_057285.3 | |||
XM_006713654.2 | 3623 | UTR 3 | XP_006713717.1 | |||
XM_011512881.1 | 3623 | UTR 3 | XP_011511183.1 | |||
XM_017006580.1 | 3623 | UTR 3 | XP_016862069.1 | |||
XM_017006581.1 | 3623 | UTR 3 | XP_016862070.1 | |||
XM_017006582.1 | 3623 | UTR 3 | XP_016862071.1 |
RPL39P5 - ribosomal protein L39 pseudogene 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |