Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGCCAGGCTGAGGACCATCCAATC[A/G]TATCCCCTTATTTTGCCAAAGAGAA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 602925 | |||||||||||||||||||||||
Literature Links: |
PIK3CB PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
PIK3CB - phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256045.1 | Intron | NP_001242974.1 | ||||
NM_006219.2 | Intron | NP_006210.1 | ||||
XM_005247530.2 | Intron | XP_005247587.1 | ||||
XM_006713659.3 | Intron | XP_006713722.1 | ||||
XM_011512895.2 | Intron | XP_011511197.1 | ||||
XM_011512896.1 | Intron | XP_011511198.1 | ||||
XM_017006619.1 | Intron | XP_016862108.1 | ||||
XM_017006620.1 | Intron | XP_016862109.1 | ||||
XM_017006621.1 | Intron | XP_016862110.1 |