Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATACTAATGCTCTGCGTGTTCCGAG[A/G]GAGAATATGGAGGCCATTTCACCTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604958 | ||||||||||||||||||||
Literature Links: |
ACTL6A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACTL6A - actin like 6A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004301.4 | 498 | Silent Mutation | AGA,AGG | R,R 79 | NP_004292.1 | |
NM_177989.3 | 498 | Silent Mutation | AGA,AGG | R,R 37 | NP_817126.1 | |
NM_178042.3 | 498 | Silent Mutation | AGA,AGG | R,R 37 | NP_829888.1 |