Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAAGGTATAACCCACTGCTTGAAC[C/T]TGTGCCTGCCAATAAAGCAAAAAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604958 MIM: 611852 | ||||||||||||||||||||
Literature Links: |
ACTL6A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACTL6A - actin like 6A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MRPL47 - mitochondrial ribosomal protein L47 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020409.2 | 415 | Missense Mutation | AAG,AGG | K,R 180 | NP_065142.2 | |
NM_177988.1 | 415 | Missense Mutation | AAG,AGG | K,R 70 | NP_817125.1 |