Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGGAGTAGTTGAATTCCTTCTAG[A/G]AAAACTAGGAATAGATGAATCTAAT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 613798 | |||||||||||||||||||||||
Literature Links: |
CCDC39 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CCDC39 - coiled-coil domain containing 39 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TTC14 - tetratricopeptide repeat domain 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042601.2 | 935 | Missense Mutation | GAA,GGA | E,G 268 | NP_001036066.1 | |
NM_001288582.1 | 935 | Missense Mutation | GAA,GGA | E,G 268 | NP_001275511.1 | |
NM_133462.3 | 935 | Missense Mutation | GAA,GGA | E,G 268 | NP_597719.1 | |
XM_011512439.2 | 935 | Missense Mutation | GAA,GGA | E,G 18 | XP_011510741.1 | |
XM_017005739.1 | 935 | Intron | XP_016861228.1 | |||
XM_017005740.1 | 935 | Missense Mutation | GAA,GGA | E,G 268 | XP_016861229.1 |