Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTCTGCTGTGCCAGAACATCCTGC[C/T]ATGTCCTGCCTGAGTCCGAGGGTTC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 614214 | |||||||||||||||||||||||
Literature Links: |
KLHL6 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
KLHL6 - kelch like family member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_130446.2 | 4920 | UTR 3 | NP_569713.2 | |||
XM_011513273.2 | 4920 | UTR 3 | XP_011511575.1 | |||
XM_011513274.2 | 4920 | Intron | XP_011511576.1 |