Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCTAGACCTGATCAATAAAAGGC[A/G]ACGGGATGGCTACCTTTTCCAATTG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 142640 | |||||||||||||||||||||||
Literature Links: |
HRG PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
HRG - histidine rich glycoprotein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000412.4 | 205 | Missense Mutation | CAA,CGA | Q,R 42 | NP_000403.1 | |
XM_005247415.3 | 205 | Missense Mutation | CAA,CGA | Q,R 42 | XP_005247472.1 |