Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAATGCAACCTAAATATCCTAGCA[C/T]TCATCTGGGTTGGAATTGCAAAGGG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
TPRG1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
TPRG1 - tumor protein p63 regulated 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198485.3 | Intron | NP_940887.1 | ||||
XM_005247382.4 | Intron | XP_005247439.1 | ||||
XM_005247388.3 | Intron | XP_005247445.1 | ||||
XM_011512732.2 | Intron | XP_011511034.2 | ||||
XM_011512734.2 | Intron | XP_011511036.1 | ||||
XM_011512735.2 | Intron | XP_011511037.1 | ||||
XM_011512736.2 | Intron | XP_011511038.1 | ||||
XM_017006257.1 | Intron | XP_016861746.1 | ||||
XM_017006258.1 | Intron | XP_016861747.1 | ||||
XM_017006259.1 | Intron | XP_016861748.1 | ||||
XM_017006260.1 | Intron | XP_016861749.1 | ||||
XM_017006261.1 | Intron | XP_016861750.1 | ||||
XM_017006262.1 | Intron | XP_016861751.1 | ||||
XM_017006263.1 | Intron | XP_016861752.1 |