Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTGTTCATGGCTTTCATAGCCTC[C/T]GTTGATGTAGAATCCATTTTTGCCA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 606487 | |||||||||||||||||||||||
Literature Links: |
ATP13A5 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ATP13A5 - ATPase 13A5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198505.2 | 3568 | Missense Mutation | AGA,GGA | R,G 1190 | NP_940907.2 | |
XM_011512770.1 | 3568 | Intron | XP_011511072.1 | |||
XM_017006304.1 | 3568 | Missense Mutation | AGA,GGA | R,G 772 | XP_016861793.1 | |
XM_017006305.1 | 3568 | Missense Mutation | AGA,GGA | R,G 631 | XP_016861794.1 |
HRASLS - HRAS like suppressor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020386.4 | 3568 | Intron | NP_065119.2 | |||
XM_011513034.1 | 3568 | Intron | XP_011511336.1 | |||
XM_011513035.2 | 3568 | Intron | XP_011511337.1 | |||
XM_011513037.2 | 3568 | Intron | XP_011511339.1 | |||
XM_017006923.1 | 3568 | Intron | XP_016862412.1 |