Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGGCCCCAGTCTCCCCTGCATTT[C/T]TTCCCTGATTGATTGATAGGAGGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607645 MIM: 606046 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NSG1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NSG1 - neuron specific gene family member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040101.1 | 1724 | Intron | NP_001035190.1 | |||
NM_001287763.1 | 1724 | Intron | NP_001274692.1 | |||
NM_001287764.1 | 1724 | Intron | NP_001274693.1 | |||
NM_014392.4 | 1724 | Intron | NP_055207.1 | |||
XM_017008022.1 | 1724 | Intron | XP_016863511.1 |
STX18 - syntaxin 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016930.2 | 1724 | UTR 3 | NP_058626.1 | |||
XM_011513480.1 | 1724 | Intron | XP_011511782.1 | |||
XM_011513483.1 | 1724 | Intron | XP_011511785.1 | |||
XM_011513485.2 | 1724 | Intron | XP_011511787.1 | |||
XM_017008296.1 | 1724 | Intron | XP_016863785.1 | |||
XM_017008297.1 | 1724 | Intron | XP_016863786.1 | |||
XM_017008298.1 | 1724 | Intron | XP_016863787.1 |