Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCAGCACGCTCCTGCTAGCCCTGG[A/G]ACCCACTGTGAGCCACTGCGGGGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 134934 MIM: 604407 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FGFR3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FGFR3 - fibroblast growth factor receptor 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LETM1 - leucine zipper and EF-hand containing transmembrane protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012318.2 | 4953 | UTR 3 | NP_036450.1 | |||
XM_006713884.1 | 4953 | UTR 3 | XP_006713947.1 | |||
XM_017008187.1 | 4953 | Intron | XP_016863676.1 |