Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGTCCATGTGACCAAAGCCGCACC[C/T]GCTTTATATGTGACTCAGAGCCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605997 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPP2R2C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PPP2R2C - protein phosphatase 2 regulatory subunit Bgamma | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206994.1 | Intron | NP_001193923.1 | ||||
NM_001206995.1 | Intron | NP_001193924.1 | ||||
NM_001206996.1 | Intron | NP_001193925.1 | ||||
NM_020416.3 | Intron | NP_065149.2 | ||||
NM_181876.2 | Intron | NP_870991.1 | ||||
XM_005247979.4 | Intron | XP_005248036.1 | ||||
XM_011513495.1 | Intron | XP_011511797.1 | ||||
XM_011513496.1 | Intron | XP_011511798.1 |