Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCAGCAAGCTTTTACCAACGTGCT[G/T]TGCTGGCAAATGCTCTTACCAGTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM33 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMEM33 - transmembrane protein 33 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018126.2 | 327 | Missense Mutation | GTG,TTG | V,L 59 | NP_060596.2 | |
XM_005248116.3 | 327 | Missense Mutation | GTG,TTG | V,L 59 | XP_005248173.1 | |
XM_005248117.2 | 327 | Missense Mutation | GTG,TTG | V,L 59 | XP_005248174.1 |