Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTCACAGTTTTAACTCTTAATTC[C/T]AAAGTAGCATCTTCATCGCATACAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613222 | ||||||||||||||||||||
Literature Links: |
GNPDA2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GNPDA2 - glucosamine-6-phosphate deaminase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270880.1 | 829 | Silent Mutation | TTA,TTG | L,L 211 | NP_001257809.1 | |
NM_001270881.1 | 829 | Silent Mutation | TTA,TTG | L,L 175 | NP_001257810.1 | |
NM_138335.2 | 829 | Silent Mutation | TTA,TTG | L,L 245 | NP_612208.1 |
GUF1 - GUF1 homolog, GTPase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |