Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATGTGTTACATAGTGACATTTCAC[A/G]AGGAAATTGCCATTGCTTTTCTGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CWH43 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CWH43 - cell wall biogenesis 43 C-terminal homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286791.1 | Intron | NP_001273720.1 | ||||
NM_025087.2 | Intron | NP_079363.2 | ||||
XM_011513755.1 | Intron | XP_011512057.1 | ||||
XM_011513756.2 | Intron | XP_011512058.1 | ||||
XM_011513757.2 | Intron | XP_011512059.1 | ||||
XM_011513758.1 | Intron | XP_011512060.1 | ||||
XM_011513759.1 | Intron | XP_011512061.1 |