Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCTGTAGCCTCTTGGTCTTTGTG[A/G]TATTTCTACAAAACAGGGACGATGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608182 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNIP4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KCNIP4 - potassium voltage-gated channel interacting protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001035003.1 | 2018 | UTR 3 | NP_001030175.1 | |||
NM_001035004.1 | 2018 | UTR 3 | NP_001030176.1 | |||
NM_025221.5 | 2018 | UTR 3 | NP_079497.2 | |||
NM_147181.3 | 2018 | UTR 3 | NP_671710.1 | |||
NM_147182.3 | 2018 | UTR 3 | NP_671711.1 | |||
NM_147183.3 | 2018 | UTR 3 | NP_671712.1 | |||
XM_005248190.4 | 2018 | UTR 3 | XP_005248247.1 | |||
XM_011513882.2 | 2018 | UTR 3 | XP_011512184.1 | |||
XM_011513883.2 | 2018 | UTR 3 | XP_011512185.1 | |||
XM_011513884.2 | 2018 | UTR 3 | XP_011512186.1 | |||
XM_011513885.2 | 2018 | UTR 3 | XP_011512187.1 | |||
XM_011513886.2 | 2018 | UTR 3 | XP_011512188.1 | |||
XM_011513887.2 | 2018 | UTR 3 | XP_011512189.1 | |||
XM_011513889.2 | 2018 | UTR 3 | XP_011512191.1 | |||
XM_017008653.1 | 2018 | UTR 3 | XP_016864142.1 |