Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCAGGACACACGTGTCGCGTATCT[A/G]TGGGGTGTTATCAGGGTTATATCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 185490 | ||||||||||||||||||||
Literature Links: |
CCDC149 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC149 - coiled-coil domain containing 149 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005248210.3 | 3889 | UTR 3 | XP_005248267.1 | |||
XM_011513906.2 | 3889 | UTR 3 | XP_011512208.1 | |||
XM_011513907.2 | 3889 | UTR 3 | XP_011512209.1 | |||
XM_011513908.1 | 3889 | UTR 3 | XP_011512210.1 | |||
XM_017008827.1 | 3889 | UTR 3 | XP_016864316.1 | |||
XM_017008828.1 | 3889 | UTR 3 | XP_016864317.1 |
SOD3 - superoxide dismutase 3, extracellular | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |