Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATCCAGGGAGCTGTCCCCGCCCC[A/G]ATCTGGCTCTCTCTGCCAGTTACAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607852 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NKD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NKD2 - naked cuticle homolog 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271082.1 | Intron | NP_001258011.1 | ||||
NM_033120.3 | Intron | NP_149111.1 | ||||
XM_005248382.3 | Intron | XP_005248439.1 | ||||
XM_005248383.3 | Intron | XP_005248440.1 | ||||
XM_011514150.2 | Intron | XP_011512452.1 | ||||
XM_017010000.1 | Intron | XP_016865489.1 |