Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAACGGCGCTGCGCGACTTGTTCT[G/T]GGAGGCGACGTCCCACCTGTTGGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615712 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
OTULIN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OTULIN - OTU deubiquitinase with linear linkage specificity | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138348.4 | Intron | NP_612357.4 | ||||
XM_011514151.2 | Intron | XP_011512453.1 | ||||
XM_011514152.2 | Intron | XP_011512454.1 | ||||
XM_011514154.2 | Intron | XP_011512456.1 | ||||
XM_017010015.1 | Intron | XP_016865504.1 |