Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTGAAGATTTAGCTTTATATCTA[A/G]TTGAGATTAACATTTGCCAAGATAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604073 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF131 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF131 - zinc finger protein 131 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005248359.2 | Intron | XP_005248416.1 | ||||
XM_005248360.2 | Intron | XP_005248417.1 | ||||
XM_005248361.2 | Intron | XP_005248418.1 | ||||
XM_005248362.4 | Intron | XP_005248419.1 | ||||
XM_005248363.4 | Intron | XP_005248420.1 | ||||
XM_005248365.3 | Intron | XP_005248422.1 | ||||
XM_005248367.2 | Intron | XP_005248424.1 | ||||
XM_011514123.2 | Intron | XP_011512425.1 | ||||
XM_011514124.2 | Intron | XP_011512426.1 | ||||
XM_017009830.1 | Intron | XP_016865319.1 | ||||
XM_017009831.1 | Intron | XP_016865320.1 | ||||
XM_017009832.1 | Intron | XP_016865321.1 | ||||
XM_017009833.1 | Intron | XP_016865322.1 | ||||
XM_017009834.1 | Intron | XP_016865323.1 | ||||
XM_017009835.1 | Intron | XP_016865324.1 | ||||
XM_017009836.1 | Intron | XP_016865325.1 | ||||
XM_017009837.1 | Intron | XP_016865326.1 | ||||
XM_017009838.1 | Intron | XP_016865327.1 | ||||
XM_017009839.1 | Intron | XP_016865328.1 | ||||
XM_017009840.1 | Intron | XP_016865329.1 | ||||
XM_017009841.1 | Intron | XP_016865330.1 | ||||
XM_017009842.1 | Intron | XP_016865331.1 | ||||
XM_017009843.1 | Intron | XP_016865332.1 |