Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTTGGTAAAATTCATCACGGAAG[C/T]TATCTGGGCCTGTAGTTTTCTTTAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SGTB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SGTB - small glutamine rich tetratricopeptide repeat containing beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019072.2 | 4489 | UTR 3 | NP_061945.1 | |||
XM_005248548.3 | 4489 | UTR 3 | XP_005248605.1 | |||
XM_017009597.1 | 4489 | UTR 3 | XP_016865086.1 |
TRAPPC13 - trafficking protein particle complex 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001093755.1 | 4489 | Intron | NP_001087224.1 | |||
NM_001093756.1 | 4489 | Intron | NP_001087225.1 | |||
NM_001243737.1 | 4489 | Intron | NP_001230666.1 | |||
NM_024941.3 | 4489 | Intron | NP_079217.2 | |||
XM_011543652.2 | 4489 | Intron | XP_011541954.1 | |||
XM_017009890.1 | 4489 | Intron | XP_016865379.1 |