Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGCTTGTCCAGATGAACGCTTGT[C/T]ACCAAACCGAGTACAGATCGTCTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAST4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAST4 - microtubule associated serine/threonine kinase family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164664.1 | Intron | NP_001158136.1 | ||||
NM_001290226.1 | Intron | NP_001277155.1 | ||||
NM_001290227.1 | Intron | NP_001277156.1 | ||||
NM_001290228.1 | Intron | NP_001277157.1 | ||||
NM_001297651.1 | Intron | NP_001284580.1 | ||||
NM_015183.2 | Intron | NP_055998.1 | ||||
NM_198828.2 | Intron | NP_942123.1 | ||||
XM_006714606.3 | Intron | XP_006714669.1 | ||||
XM_006714610.2 | Intron | XP_006714673.1 | ||||
XM_011543382.2 | Intron | XP_011541684.1 | ||||
XM_011543384.2 | Intron | XP_011541686.1 | ||||
XM_011543385.2 | Intron | XP_011541687.1 | ||||
XM_011543386.2 | Intron | XP_011541688.1 | ||||
XM_017009447.1 | Intron | XP_016864936.1 | ||||
XM_017009448.1 | Intron | XP_016864937.1 | ||||
XM_017009449.1 | Intron | XP_016864938.1 | ||||
XM_017009450.1 | Intron | XP_016864939.1 | ||||
XM_017009451.1 | Intron | XP_016864940.1 | ||||
XM_017009452.1 | Intron | XP_016864941.1 | ||||
XM_017009453.1 | Intron | XP_016864942.1 |