Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTTCACTGGAACAAGAAAAGAAA[A/T]CTGAAAAGCCATCGACTCCAGTCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607012 | ||||||||||||||||||||
Literature Links: |
BDP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BDP1 - B double prime 1, subunit of RNA polymerase III transcription initiation factor IIIB | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018429.2 | 867 | Missense Mutation | ACT,TCT | T,S 209 | NP_060899.2 | |
XM_011543511.2 | 867 | Missense Mutation | ACT,TCT | T,S 209 | XP_011541813.1 | |
XM_011543512.2 | 867 | Missense Mutation | ACT,TCT | T,S 209 | XP_011541814.1 | |
XM_017009630.1 | 867 | Missense Mutation | ACT,TCT | T,S 209 | XP_016865119.1 | |
XM_017009631.1 | 867 | Missense Mutation | ACT,TCT | T,S 209 | XP_016865120.1 | |
XM_017009632.1 | 867 | Missense Mutation | ACT,TCT | T,S 209 | XP_016865121.1 | |
XM_017009633.1 | 867 | Missense Mutation | ACT,TCT | T,S 209 | XP_016865122.1 | |
XM_017009634.1 | 867 | Missense Mutation | ACT,TCT | T,S 209 | XP_016865123.1 |