Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTTTGGTTTTTGTCCCCACGGTT[A/G]ATCAGATACAAAGACAAGGGAGAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611989 MIM: 615484 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MRPS27 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MRPS27 - mitochondrial ribosomal protein S27 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PTCD2 - pentatricopeptide repeat domain 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284403.1 | Intron | NP_001271332.1 | ||||
NM_001284404.1 | Intron | NP_001271333.1 | ||||
NM_001284405.1 | Intron | NP_001271334.1 | ||||
NM_024754.4 | Intron | NP_079030.3 | ||||
XM_005248601.2 | Intron | XP_005248658.1 | ||||
XM_005248603.2 | Intron | XP_005248660.1 | ||||
XM_017009866.1 | Intron | XP_016865355.1 | ||||
XM_017009867.1 | Intron | XP_016865356.1 | ||||
XM_017009868.1 | Intron | XP_016865357.1 |