Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCACCCATGTTAGGTACAGCTTTG[A/C]AGGCTGTGATGAGGATATAAGATCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602901 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TNPO1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TNPO1 - transportin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002270.3 | Intron | NP_002261.3 | ||||
NM_153188.2 | Intron | NP_694858.1 | ||||
XM_005248500.2 | Intron | XP_005248557.1 | ||||
XM_005248501.1 | Intron | XP_005248558.1 | ||||
XM_017009458.1 | Intron | XP_016864947.1 |