Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCGTCTTCTTCGTCCTCGCCCTC[C/G]CCCACCACGTCGATGTCTGTTTCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601091 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FOXD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FOXD1 - forkhead box D1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004472.2 | 234 | Silent Mutation | GGC,GGG | G,G 23 | NP_004463.1 |
LINC01386 - long intergenic non-protein coding RNA 1386 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |