Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTTGATATAAGTGGGAGGGCTCC[C/T]CTCAGGCAGTTTGAAGGCCATACAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605787 MIM: 616194 | ||||||||||||||||||||
Literature Links: |
ANKRA2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ANKRA2 - ankyrin repeat family A member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UTP15 - UTP15, small subunit processome component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284430.1 | 858 | Silent Mutation | CCC,CCT | P,P 95 | NP_001271359.1 | |
NM_001284431.1 | 858 | UTR 5 | NP_001271360.1 | |||
NM_032175.3 | 858 | Silent Mutation | CCC,CCT | P,P 114 | NP_115551.2 | |
XM_011543680.2 | 858 | Silent Mutation | CCC,CCT | P,P 114 | XP_011541982.1 |