Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCAGTCTGAGGCAGAGTGGAGATA[C/T]AGCAGAGCAAGAGGTTTGGTTGGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601919 MIM: 605401 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
F2RL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
F2RL2 - coagulation factor II thrombin receptor like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256566.1 | 2432 | UTR 3 | NP_001243495.1 | |||
NM_004101.3 | 2432 | UTR 3 | NP_004092.1 |
IQGAP2 - IQ motif containing GTPase activating protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001285460.1 | 2432 | Intron | NP_001272389.1 | |||
NM_001285461.1 | 2432 | Intron | NP_001272390.1 | |||
NM_001285462.1 | 2432 | Intron | NP_001272391.1 | |||
NM_006633.3 | 2432 | Intron | NP_006624.2 | |||
XM_005248410.2 | 2432 | Intron | XP_005248467.1 | |||
XM_005248414.1 | 2432 | Intron | XP_005248471.1 | |||
XM_011543108.1 | 2432 | Intron | XP_011541410.1 | |||
XM_017008960.1 | 2432 | Intron | XP_016864449.1 |
LOC101929109 - uncharacterized LOC101929109 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |