Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAATTGTTTGCGATGAGATATGA[C/T]TTCACATACCATTATGTAATTTGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 187930 MIM: 605401 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
F2R PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
F2R - coagulation factor II thrombin receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001311313.1 | Intron | NP_001298242.1 | ||||
NM_001992.4 | Intron | NP_001983.2 |
IQGAP2 - IQ motif containing GTPase activating protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NCRUPAR - non-protein coding RNA, upstream of F2R/PAR1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |