Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGCCATTTAGACTATCTCTTTGC[C/T]AATTTTTGCTTACTGCTGTAGGGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603401 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AP3B1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AP3B1 - adaptor related protein complex 3 beta 1 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271769.1 | 3628 | UTR 3 | NP_001258698.1 | |||
NM_003664.4 | 3628 | UTR 3 | NP_003655.3 | |||
XM_005248618.3 | 3628 | UTR 3 | XP_005248675.1 | |||
XM_005248619.4 | 3628 | Intron | XP_005248676.1 | |||
XM_017010001.1 | 3628 | UTR 3 | XP_016865490.1 |