Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTTTCTCCACTCATGCCACTGCC[A/G]GCTGATTGGCATGACTGCGTTCCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARL15 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARL15 - ADP ribosylation factor like GTPase 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019087.2 | Intron | NP_061960.1 | ||||
XM_011543498.2 | Intron | XP_011541800.1 | ||||
XM_011543499.2 | Intron | XP_011541801.1 | ||||
XM_011543500.2 | Intron | XP_011541802.1 | ||||
XM_017009598.1 | Intron | XP_016865087.1 |