Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTTTCTGTTTCCAAGCCTGGCTG[A/T]CTGGTCTTTCAGCTAAGTCTATGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606457 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IBTK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IBTK - inhibitor of Bruton tyrosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300906.1 | Intron | NP_001287835.1 | ||||
NM_015525.3 | Intron | NP_056340.2 | ||||
XM_006715453.2 | Intron | XP_006715516.1 | ||||
XM_006715454.2 | Intron | XP_006715517.1 |