Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCCAACGGTTTTTCTCGACTTCT[A/G]ATAAGAGCTGCTTCTTGTTGTAGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606457 | ||||||||||||||||||||
Literature Links: |
IBTK PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IBTK - inhibitor of Bruton tyrosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300906.1 | 5110 | Silent Mutation | NP_001287835.1 | |||
NM_015525.3 | 5110 | Silent Mutation | NP_056340.2 | |||
XM_006715453.2 | 5110 | Silent Mutation | XP_006715516.1 | |||
XM_006715454.2 | 5110 | Silent Mutation | XP_006715517.1 |