Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACACAATACACAAGCATGAGGTGAT[A/G]ACCAAGAAGAGGATGAAGATCGTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611921 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GJB7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GJB7 - gap junction protein beta 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198568.2 | 898 | Silent Mutation | GTC,GTT | V,V 184 | NP_940970.1 | |
XM_005248716.2 | 898 | Silent Mutation | GTC,GTT | V,V 184 | XP_005248773.1 |