Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTAGAAGAAAAAAATGGTAGTAG[C/T]GGAAGCAGTAGATTGCTGCCACAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611921 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C6orf163 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C6orf163 - chromosome 6 open reading frame 163 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GJB7 - gap junction protein beta 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SMIM8 - small integral membrane protein 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042493.2 | Intron | NP_001035958.1 | ||||
NM_001287445.1 | Intron | NP_001274374.1 | ||||
NM_020425.5 | Intron | NP_065158.3 |