Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATCGCTGAGTGTCTCAGGAAGTGC[A/G]TTTACATCTCGAAGAATGATATAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101929057 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC101929057 - uncharacterized LOC101929057 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LYRM2 - LYR motif containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MDN1 - midasin AAA ATPase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014611.2 | 16711 | Silent Mutation | AAC,AAT | N,N 5573 | NP_055426.1 | |
XM_005248700.3 | 16711 | Silent Mutation | AAC,AAT | N,N 3568 | XP_005248757.1 | |
XM_006715405.3 | 16711 | Silent Mutation | AAC,AAT | N,N 3721 | XP_006715468.1 | |
XM_011535635.2 | 16711 | Silent Mutation | AAC,AAT | N,N 5476 | XP_011533937.1 | |
XM_011535636.2 | 16711 | Intron | XP_011533938.1 |