Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGAAAATAACTAGCTTACTGAAAT[G/T]CTTGTCTTTTAAAGAAAGTTGGGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614348 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADTRP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADTRP - androgen dependent TFPI regulating protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143948.1 | 1563 | UTR 3 | NP_001137420.1 | |||
NM_032744.3 | 1563 | UTR 3 | NP_116133.1 | |||
XM_005249454.3 | 1563 | UTR 3 | XP_005249511.1 | |||
XM_011514956.1 | 1563 | UTR 3 | XP_011513258.1 | |||
XM_011514958.2 | 1563 | Intron | XP_011513260.1 | |||
XM_011514959.1 | 1563 | Intron | XP_011513261.1 | |||
XM_017011377.1 | 1563 | UTR 3 | XP_016866866.1 |
LOC340184 - uncharacterized LOC340184 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |