Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTTCCTGAAGGAATTTCTTACTT[C/T]TTCCCATTGTGTCATTTATAGAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602884 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GMDS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GMDS - GDP-mannose 4,6-dehydratase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001253846.1 | Intron | NP_001240775.1 | ||||
NM_001500.3 | Intron | NP_001491.1 | ||||
XM_006715066.2 | Intron | XP_006715129.1 | ||||
XM_011514500.1 | Intron | XP_011512802.1 | ||||
XM_011514502.2 | Intron | XP_011512804.1 | ||||
XM_011514503.2 | Intron | XP_011512805.1 | ||||
XM_011514505.1 | Intron | XP_011512807.1 | ||||
XM_011514506.1 | Intron | XP_011512808.1 | ||||
XM_011514507.1 | Intron | XP_011512809.1 | ||||
XM_017010752.1 | Intron | XP_016866241.1 |