Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGAAAAGAGACTGTGTGAGCATT[C/T]TCACACCAGGAAATGGGCTGAGAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
HMGN4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HMGN4 - high mobility group nucleosomal binding domain 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006353.2 | 163 | Intron | NP_006344.1 | |||
XM_017010171.1 | 163 | UTR 5 | XP_016865660.1 | |||
XM_017010172.1 | 163 | Intron | XP_016865661.1 | |||
XM_017010173.1 | 163 | UTR 5 | XP_016865662.1 |