Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTGAACTCTGGGGTTCCATTCCT[C/T]ACCTCCTTCCCTGGAACCACCTCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 109170 MIM: 600978 MIM: 611550 | ||||||||||||||||||||
Literature Links: |
LST1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LST1 - leukocyte specific transcript 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LTB - lymphotoxin beta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NCR3 - natural cytotoxicity triggering receptor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145466.1 | 408 | Silent Mutation | GTA,GTG | V,V 66 | NP_001138938.1 | |
NM_001145467.1 | 408 | Silent Mutation | GTA,GTG | V,V 66 | NP_001138939.1 | |
NM_147130.2 | 408 | Silent Mutation | GTA,GTG | V,V 66 | NP_667341.1 | |
XM_006715049.3 | 408 | Silent Mutation | GTA,GTG | V,V 66 | XP_006715112.1 | |
XM_011514459.2 | 408 | Intron | XP_011512761.1 |