Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCTTAAAGTCTTTACCCAGCCCC[A/G]GGATGCGACCGGAGAAAAAAAAGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608272 MIM: 606107 | ||||||||||||||||||||
Literature Links: |
NEU1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NEU1 - neuraminidase 1 (lysosomal sialidase) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC44A4 - solute carrier family 44 member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001178044.1 | 1937 | Missense Mutation | NP_001171515.1 | |||
NM_001178045.1 | 1937 | Missense Mutation | NP_001171516.1 | |||
NM_025257.2 | 1937 | Missense Mutation | NP_079533.2 |