Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTCCGCATCCCCTGGAAGCACGC[G/T]GGCAAGCAGGACTACAACCGCGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601900 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IRF4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IRF4 - interferon regulatory factor 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195286.1 | 297 | Silent Mutation | GCG,GCT | A,A 57 | NP_001182215.1 | |
NM_002460.3 | 297 | Silent Mutation | GCG,GCT | A,A 57 | NP_002451.2 | |
XM_006715090.2 | 297 | Silent Mutation | GCG,GCT | A,A 57 | XP_006715153.1 |