Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCACAGCTCGGTCCCAGACACGCT[C/T]CATGCGCCTCCTGCAATGAGAGGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616312 | ||||||||||||||||||||
Literature Links: |
LEMD2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LEMD2 - LEM domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143944.1 | 804 | Missense Mutation | AAG,GAG | K,E 156 | NP_001137416.1 | |
NM_181336.3 | 804 | Missense Mutation | AAG,GAG | K,E 458 | NP_851853.1 | |
XM_017010437.1 | 804 | Intron | XP_016865926.1 |