Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGCAAATTCTAATTTACTGGCAT[A/G]AATTGTACAAAATAACTGCTCATCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615932 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCTD20 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KCTD20 - potassium channel tetramerization domain containing 20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286579.1 | Intron | NP_001273508.1 | ||||
NM_001286580.1 | Intron | NP_001273509.1 | ||||
NM_173562.4 | Intron | NP_775833.2 | ||||
XM_005248940.4 | Intron | XP_005248997.1 | ||||
XM_006715023.2 | Intron | XP_006715086.1 | ||||
XM_011514398.2 | Intron | XP_011512700.1 |
PXT1 - peroxisomal, testis specific 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |