Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTAAAGGCTCCCTCCAGACACGA[C/T]TTTTCCTTTGTCACCGTACTCCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
PI16 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PI16 - peptidase inhibitor 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199159.1 | Intron | NP_001186088.1 | ||||
NM_153370.2 | Intron | NP_699201.2 | ||||
XM_005248917.2 | Intron | XP_005248974.1 | ||||
XM_011514375.2 | Intron | XP_011512677.1 | ||||
XM_017010430.1 | Intron | XP_016865919.1 |