Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTCATTTTCTTTAGGGAAATAGAT[C/T]TTGTTAGTATTTCATTTTTATAATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607455 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZFAND3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZFAND3 - zinc finger AN1-type containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021943.2 | Intron | NP_068762.1 | ||||
XM_005249269.1 | Intron | XP_005249326.1 | ||||
XM_011514790.1 | Intron | XP_011513092.1 | ||||
XM_017011171.1 | Intron | XP_016866660.1 |