Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTGGACAGAAATGTAGAAAAACA[A/G]GAAGGCAGAAGATGAGCAAACGGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TSPO2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TSPO2 - translocator protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010873.2 | Intron | NP_001010873.1 | ||||
NM_001159726.1 | Intron | NP_001153198.1 | ||||
XM_011514396.2 | Intron | XP_011512698.1 | ||||
XM_011514397.2 | Intron | XP_011512699.1 |
UNC5CL - unc-5 family C-terminal like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |