Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGTCAGTGACTCCATTCTTCAGA[C/T]AATGAGACTTCATTGCCTCTAAACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604797 MIM: 189903 MIM: 614393 | ||||||||||||||||||||
Literature Links: |
APOBEC2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
APOBEC2 - apolipoprotein B mRNA editing enzyme catalytic subunit 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NFYA - nuclear transcription factor Y subunit alpha | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OARD1 - O-acyl-ADP-ribose deacylase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |